Assay Details
| Gene Symbol: SFT2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053457.1 | XP_017908946.1 | 6 - 7 | 496 | 69 | |
| XM_018053458.1 | XP_017908947.1 | 6 - 7 | 495 | 69 | ||
More Information
Set Membership: |
|

SFT2D1
Genomic Map