Assay Details
| Gene Symbol: TMEM255B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044983.1 | XP_017900472.1 | 4 - 5 | 460 | 59 | |
| XM_018044984.1 | XP_017900473.1 | 3 - 4 | 244 | 59 | ||
More Information
Set Membership: |
|

TMEM255B
Genomic Map