Assay Details
| Gene Symbol: ZNF704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058379.1 | XP_017913868.1 | 6 - 7 | 1055 | 66 | |
| XM_018058380.1 | XP_017913869.1 | 6 - 7 | 1203 | 66 | ||
| GenBank mRNA | JO545123.2 | - | 3 - 4 | 470 | 66 | |
More Information
Set Membership: |
|

ZNF704
Genomic Map