Assay Details
| Gene Symbol: PECR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059757.1 | XP_017915246.1 | 7 - 8 | 892 | 62 | |
| GenBank mRNA | JO437342.2 | - | 1 - 2 | 344 | 62 | |
More Information
Set Membership: |
|

PECR
Genomic Map