Assay Details
| Gene Symbol: TTLL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972587.2 | XP_013828041.1 | 9 - 10 | 1895 | 60 | |
| XM_013972588.2 | XP_013828042.1 | 7 - 8 | 1234 | 60 | ||
| XM_013972589.2 | XP_013828043.1 | 6 - 7 | 778 | 60 | ||
| XM_018064928.1 | XP_017920417.1 | 8 - 9 | 1297 | 60 | ||
More Information
Set Membership: |
|

TTLL6
Genomic Map