Assay Details
| Gene Symbol: PPIL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039213.1 | XP_017894702.1 | 3 - 4 | 558 | 86 | |
| GenBank mRNA | JO612386.2 | - | 1 - 2 | 1123 | 86 | |
More Information
Set Membership: |
|

PPIL1
Genomic Map