Assay Details
| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038550.1 | XP_017894039.1 | 4 - 5 | 1079 | 64 | |
| GenBank mRNA | JI861632.1 | - | 3 - 4 | 646 | 64 | |
More Information
Set Membership: |
|

ZKSCAN8
Genomic Map