Assay Details
| Gene Symbol: LOC102171580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976156.2 | XP_013831610.2 | 3 - 4 | 446 | 80 | |
| GenBank mRNA | JO570095.2 | - | 3 - 4 | 373 | 80 | |
More Information
Set Membership: |
|

LOC102171580
Genomic Map