Assay Details
Gene Symbol: KIRREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046339.1 | XP_017901828.1 | 11 - 12 | 1871 | 57 | |
XM_018046340.1 | XP_017901829.1 | 11 - 12 | 1822 | 57 | ||
XM_018046341.1 | XP_017901830.1 | 10 - 11 | 1723 | 57 | ||
XM_018046342.1 | XP_017901831.1 | 11 - 12 | 2541 | 57 | ||
GenBank mRNA | JO369516.2 | - | 3 - 4 | 346 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
