Assay Details
| Gene Symbol: AP2M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675196.3 | XP_005675253.1 | 10 - 11 | 1223 | 66 | |
| XM_005675197.3 | XP_005675254.1 | 9 - 10 | 1217 | 66 | ||
| XM_013964104.2 | XP_013819558.1 | 9 - 10 | 1195 | 66 | ||
| XM_018047286.1 | XP_017902775.1 | 10 - 11 | 1305 | 66 | ||
More Information
Set Membership: |
|

AP2M1
Genomic Map