Assay Details
| Gene Symbol: CD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677813.3 | XP_005677870.1 | 3 - 4 | 755 | 85 | |
| XM_013962501.2 | XP_013817955.2 | 3 - 4 | 1076 | 85 | ||
| GenBank mRNA | JO580411.1 | - | 2 - 3 | 1209 | 85 | |
More Information
Set Membership: |
|

CD2
Genomic Map