Assay Details
| Gene Symbol: COLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684369.3 | XP_005684426.3 | 2 - 3 | 416 | 63 | |
| XM_018052717.1 | XP_017908206.1 | 2 - 3 | 415 | 63 | ||
| XM_018052718.1 | XP_017908207.1 | 2 - 3 | 515 | 63 | ||
| GenBank mRNA | JO419983.2 | - | 5 - 6 | 1040 | 63 | |
| JO436490.2 | - | 5 - 6 | 1058 | 63 | ||
More Information
Set Membership: |
|

COLEC11
Genomic Map