Assay Details
| Gene Symbol: LRAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061625.1 | XP_017917114.1 | 2 - 3 | 590 | 80 | |
| GenBank mRNA | JO395905.2 | - | 1 - 2 | 447 | 80 | |
More Information
Set Membership: |
|

LRAT
Genomic Map