Assay Details
Gene Symbol: TNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064571.1 | XP_017920060.1 | 4 - 5 | 511 | 62 | |
XM_018064572.1 | XP_017920061.1 | 4 - 5 | 511 | 62 | ||
XM_018064573.1 | XP_017920062.1 | 3 - 4 | 419 | 62 | ||
GenBank mRNA | JT676101.1 | - | 4 - 5 | 470 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
