Assay Details
| Gene Symbol: ELOVL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698356.2 | XP_005698413.1 | 3 - 4 | 579 | 65 | |
| GenBank mRNA | KA351780.1 | - | 3 - 4 | 617 | 65 | |
More Information
Set Membership: |
|

ELOVL3
Genomic Map