Assay Details
| Gene Symbol: CAMLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965572.2 | XP_013821026.1 | 2 - 3 | 868 | 81 | |
| GenBank mRNA | KA312819.1 | - | 2 - 3 | 658 | 81 | |
More Information
Set Membership: |
|

CAMLG
Genomic Map