Assay Details
| Gene Symbol: DUOXA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054141.1 | XP_017909630.1 | 2 - 3 | 642 | 73 | |
| XM_018054142.1 | XP_017909631.1 | 3 - 4 | 586 | 73 | ||
| GenBank mRNA | KA311078.1 | - | 3 - 4 | 213 | 73 | |
More Information
Set Membership: |
|

DUOXA2
Genomic Map