Assay Details
| Gene Symbol: LRRN4CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042989.1 | XP_017898478.1 | 1 - 2 | 781 | 74 | |
| GenBank mRNA | JO633491.2 | - | 1 - 2 | 265 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018042989.1 transcripts.
Set Membership: |
|

LRRN4CL
Genomic Map