Assay Details
| Gene Symbol: HRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043553.1 | XP_017899042.1 | 3 - 4 | 462 | 68 | |
| XM_018043554.1 | XP_017899043.1 | 3 - 4 | 354 | 68 | ||
| XM_018043555.1 | XP_017899044.1 | 3 - 4 | 462 | 68 | ||
| GenBank mRNA | JO366446.2 | - | 3 - 4 | 395 | 68 | |
More Information
Set Membership: |
|

HRAS
Genomic Map