Assay Details
| Gene Symbol: CTDSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680257.3 | XP_005680314.1 | 3 - 4 | 775 | 59 | |
| GenBank mRNA | JT652429.1 | - | 4 - 5 | 745 | 59 | |
More Information
Set Membership: |
|

CTDSP2
Genomic Map