Assay Details
| Gene Symbol: PEBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684000.3 | XP_005684057.1 | 6 - 7 | 776 | 68 | |
| GenBank mRNA | JO388148.2 | - | 6 - 7 | 730 | 68 | |
| JO546860.2 | - | 3 - 4 | 223 | 68 | ||
More Information
Set Membership: |
|

PEBP4
Genomic Map