Assay Details
| Gene Symbol: GNAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039549.1 | XP_017895038.1 | 6 - 7 | 692 | 64 | |
| XM_018039550.1 | XP_017895039.1 | 5 - 6 | 1265 | 64 | ||
| GenBank mRNA | KA335280.1 | - | 5 - 6 | 420 | 64 | |
More Information
Set Membership: |
|

GNAL
Genomic Map