Assay Details
| Gene Symbol: ERCC6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700652.3 | XP_005700709.2 | 1 - 2 | 110 | 77 | |
| GenBank mRNA | JO606842.2 | - | 1 - 2 | 845 | 77 | |
More Information
Set Membership: |
|

ERCC6L
Genomic Map