Assay Details
| Gene Symbol: DESI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048825.1 | XP_017904314.1 | 5 - 6 | 646 | 79 | |
| GenBank mRNA | JO599835.2 | - | 5 - 6 | 650 | 79 | |
More Information
Set Membership: |
|

DESI1
Genomic Map