Assay Details
| Gene Symbol: PLPPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051115.1 | XP_017906604.1 | 6 - 7 | 1107 | 57 | |
| XM_018051116.1 | XP_017906605.1 | 5 - 6 | 1989 | 57 | ||
| XM_018051117.1 | XP_017906606.1 | 6 - 7 | 943 | 57 | ||
| XM_018051118.1 | XP_017906607.1 | 5 - 6 | 918 | 57 | ||
| XM_018051119.1 | XP_017906608.1 | 6 - 7 | 1108 | 57 | ||
| GenBank mRNA | JO645563.2 | - | 4 - 5 | 1502 | 57 | |
More Information
Set Membership: |
|

PLPPR2
Genomic Map