Assay Details
| Gene Symbol: OTUB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066511.1 | XP_017922000.1 | 4 - 5 | 510 | 65 | |
| GenBank mRNA | KA332420.1 | - | 2 - 3 | 3182 | 65 | |
More Information
Set Membership: |
|

OTUB2
Genomic Map