Assay Details
| Gene Symbol: SLC27A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685790.3 | XP_005685847.2 | 9 - 10 | 1870 | 75 | |
| GenBank mRNA | JO427913.1 | - | 9 - 10 | 2121 | 75 | |
More Information
Set Membership: |
|

SLC27A2
Genomic Map