Assay Details
| Gene Symbol: MFRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689544.3 | XP_005689601.2 | 8 - 9 | 1161 | 62 | |
| GenBank mRNA | JO640525.2 | - | 4 - 5 | 488 | 62 | |
More Information
Set Membership: |
|

MFRP
Genomic Map