Assay Details
| Gene Symbol: MMRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042186.1 | XP_017897675.1 | 2 - 3 | 366 | 60 | |
| GenBank mRNA | JO415925.2 | - | 2 - 3 | 369 | 60 | |
More Information
Set Membership: |
|

MMRN2
Genomic Map