Assay Details
| Gene Symbol: BSND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013962801.2 | XP_013818255.2 | 1 - 2 | 398 | 63 | |
| GenBank mRNA | JO427417.1 | - | 1 - 2 | 466 | 63 | |
| JO428493.1 | - | 1 - 2 | 466 | 63 | ||
More Information
Set Membership: |
|

BSND
Genomic Map