Assay Details
| Gene Symbol: MOB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049551.1 | XP_017905040.1 | 3 - 4 | 324 | 64 | |
| XM_018049552.1 | XP_017905041.1 | 3 - 4 | 324 | 64 | ||
| XM_018049553.1 | XP_017905042.1 | 3 - 4 | 566 | 64 | ||
| XM_018049554.1 | XP_017905043.1 | 4 - 5 | 594 | 64 | ||
| XM_018049555.1 | XP_017905044.1 | 4 - 5 | 454 | 64 | ||
| XM_018049556.1 | XP_017905045.1 | 4 - 5 | 380 | 64 | ||
| GenBank mRNA | JO506044.2 | - | 3 - 4 | 447 | 64 | |
| KA352293.1 | - | 4 - 5 | 449 | 64 | ||
More Information
Set Membership: |
|

MOB1B
Genomic Map