Assay Details
| Gene Symbol: LOC102173969 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972659.2 | XP_013828113.2 | 5 - 6 | 977 | 62 | |
| XM_013972661.2 | XP_013828115.2 | 5 - 6 | 902 | 62 | ||
| GenBank mRNA | JO324854.2 | - | 2 - 3 | 1274 | 62 | |
More Information
Set Membership: |
|

LOC102173969
Genomic Map