Assay Details
| Gene Symbol: LOC102173969 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972659.2 | XP_013828113.2 | 6 - 7 | 1086 | 83 | |
| XM_013972661.2 | XP_013828115.2 | 6 - 7 | 1011 | 83 | ||
| GenBank mRNA | JO324854.2 | - | 1 - 2 | 1165 | 83 | |
More Information
Set Membership: |
|

LOC102173969
Genomic Map