Assay Details
| Gene Symbol: PCNX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042840.1 | XP_017898329.1 | 21 - 22 | 3585 | 58 | |
| XM_018042841.1 | XP_017898330.1 | 21 - 22 | 3582 | 58 | ||
| XM_018042842.1 | XP_017898331.1 | 21 - 22 | 3570 | 58 | ||
| XM_018042843.1 | XP_017898332.1 | 21 - 22 | 3567 | 58 | ||
| XM_018042844.1 | XP_017898333.1 | 21 - 22 | 3546 | 58 | ||
| XM_018042845.1 | XP_017898334.1 | 21 - 22 | 3477 | 58 | ||
More Information
Set Membership: |
|

PCNX3
Genomic Map