Assay Details
| Gene Symbol: MAP7D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044250.1 | XP_017899739.1 | 14 - 15 | 3083 | 63 | |
| GenBank mRNA | JO354657.2 | - | 3 - 4 | 665 | 63 | |
More Information
Set Membership: |
|

MAP7D3
Genomic Map