Assay Details
| Gene Symbol: RDH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680398.3 | XP_005680455.1 | 3 - 4 | 1344 | 59 | |
| XM_013963965.2 | XP_013819419.1 | 3 - 4 | 1439 | 59 | ||
| GenBank mRNA | JO454228.2 | - | 2 - 3 | 495 | 59 | |
More Information
Set Membership: |
|

RDH5
Genomic Map