Assay Details
Gene Symbol: PPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685667.3 | XP_005685724.1 | 4 - 5 | 619 | 83 | |
GenBank mRNA | JO439798.2 | - | 1 - 2 | 324 | 83 | |
JT679567.1 | - | 4 - 5 | 637 | 83 | ||
KM076929.1 | - | 4 - 5 | 528 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
