Assay Details
| Gene Symbol: TIMM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693300.3 | XP_005693357.1 | 3 - 4 | 743 | 65 | |
| GenBank mRNA | KA319093.1 | - | 1 - 2 | 1667 | 65 | |
More Information
Set Membership: |
|

TIMM22
Genomic Map