Assay Details
| Gene Symbol: LGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698230.3 | XP_005698287.1 | 1 - 2 | 566 | 75 | |
| GenBank mRNA | JT729177.1 | - | 3 - 4 | 620 | 75 | |
More Information
Set Membership: |
|

LGI1
Genomic Map