Assay Details
| Gene Symbol: ZNF268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061460.1 | XP_017916949.1 | 4 - 5 | 967 | 67 | |
| GenBank mRNA | JT696714.1 | - | 5 - 6 | 331 | 67 | |
More Information
Set Membership: |
|

ZNF268
Genomic Map