Assay Details
| Gene Symbol: CDC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055357.1 | XP_017910846.1 | 3 - 4 | 321 | 67 | |
| XM_018055366.1 | XP_017910855.1 | 4 - 5 | 306 | 67 | ||
| XM_018055370.1 | XP_017910859.1 | 3 - 4 | 321 | 67 | ||
| GenBank mRNA | JO422138.2 | - | 3 - 4 | 691 | 67 | |
| JO579502.1 | - | 3 - 4 | 415 | 67 | ||
More Information
Set Membership: |
|

CDC42
Genomic Map