Assay Details
| Gene Symbol: P3H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065098.1 | XP_017920587.1 | 6 - 7 | 1308 | 63 | |
| GenBank mRNA | JO636690.2 | - | 6 - 7 | 955 | 63 | |
| KA311477.1 | - | 6 - 7 | 879 | 63 | ||
More Information
Set Membership: |
|

P3H4
Genomic Map