Assay Details
| Gene Symbol: TAFA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013962526.2 | XP_013817980.2 | 3 - 4 | 473 | 78 | |
| XM_018045926.1 | XP_017901415.1 | 4 - 5 | 428 | 78 | ||
More Information
Set Membership: |
|

TAFA3
Genomic Map