Assay Details
| Gene Symbol: LRRC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963247.2 | XP_013818701.2 | 3 - 4 | 1379 | 102 | |
| GenBank mRNA | JO498515.2 | - | 1 - 2 | 667 | 102 | |
| JT756247.1 | - | 3 - 4 | 1242 | 102 | ||
| KA349294.1 | - | 1 - 2 | 83 | 102 | ||
More Information
Set Membership: |
|

LRRC17
Genomic Map