Assay Details
| Gene Symbol: GMFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054124.1 | XP_017909613.1 | 3 - 4 | 247 | 75 | |
| XM_018054125.1 | XP_017909614.1 | 3 - 4 | 247 | 75 | ||
| GenBank mRNA | JO321316.2 | - | 3 - 4 | 195 | 75 | |
| JO628854.2 | - | 4 - 5 | 263 | 75 | ||
More Information
Set Membership: |
|

GMFB
Genomic Map