Assay Details
| Gene Symbol: MTFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060981.1 | XP_017916470.1 | 1 - 2 | 207 | 80 | |
| XM_018060982.1 | XP_017916471.1 | 1 - 2 | 207 | 80 | ||
| GenBank mRNA | JO641020.2 | - | 1 - 2 | 167 | 80 | |
More Information
Set Membership: |
|

MTFP1
Genomic Map