Assay Details
| Gene Symbol: ECI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039032.1 | XP_017894521.1 | 8 - 9 | 997 | 63 | |
| XM_018039033.1 | XP_017894522.1 | 8 - 9 | 923 | 63 | ||
| XM_018039034.1 | XP_017894523.1 | 8 - 9 | 919 | 63 | ||
| XM_018039035.1 | XP_017894524.1 | 7 - 8 | 842 | 63 | ||
| XM_018039036.1 | XP_017894525.1 | 7 - 8 | 846 | 63 | ||
| XM_018039037.1 | XP_017894526.1 | 9 - 10 | 1047 | 63 | ||
| XM_018039038.1 | XP_017894527.1 | 8 - 9 | 967 | 63 | ||
| XM_018039039.1 | XP_017894528.1 | 7 - 8 | 967 | 63 | ||
| XM_018039040.1 | XP_017894529.1 | 9 - 10 | 981 | 63 | ||
| GenBank mRNA | KA316061.1 | - | 3 - 4 | 946 | 63 | |
More Information
Set Membership: |
|

ECI2
Genomic Map