Assay Details
| Gene Symbol: PBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046428.1 | XP_017901917.1 | 4 - 5 | 1012 | 99 | |
| XM_018046429.1 | XP_017901918.1 | 4 - 5 | 1013 | 99 | ||
| XM_018046430.1 | XP_017901919.1 | 4 - 5 | 1010 | 99 | ||
| XM_018046431.1 | XP_017901920.1 | 4 - 5 | 1010 | 99 | ||
More Information
Set Membership: |
|

PBX1
Genomic Map