Assay Details
Gene Symbol: LOC102190533 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059837.1 | XP_017915326.1 | 14 - 15 | 3037 | 78 | |
XM_018059838.1 | XP_017915327.1 | 11 - 12 | 2689 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
