Assay Details
| Gene Symbol: LOC102190533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059837.1 | XP_017915326.1 | 4 - 5 | 422 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102190533| Gene Symbol: LOC102190533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059837.1 | XP_017915326.1 | 4 - 5 | 422 | 109 | |
Set Membership: |
|